Learn More
Chemglass Life Sciences ECODryBlock for 19mm OD Vials like 19mm x 65mm, 12mL Vials
SDP
Catalog No. 5000493108
Encompass_Preferred
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Catalog No. 50-004-93108 Supplier Chemglass Life Sciences Supplier No. CG1991V123
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.