Learn More
Chemglass Life Sciences ECODryBlock for 20mm Round Bottom Tubes, 21mm O.D x 48.4mm Deep Well, 8 Wells
SDP
Catalog No. 5000493088
Encompass_Preferred
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.
Catalog No. 50-004-93088 Supplier Chemglass Life Sciences Supplier No. CG1991V106
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.