Learn More
Chemglass Life Sciences ECODryBlock for 21mm OD vials like 21mm x 70mm, 16mL (4 Dram) Vials
SDP
Catalog No. 5000493105
Encompass_Preferred
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Catalog No. 50-004-93105 Supplier Chemglass Life Sciences Supplier No. CG1991V124
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.