Learn More
Chemglass Life Sciences ECODryBlock for 23mm OD vials like 23mm x 85mm, 24mL (6 Dram) Vials
SDP
Catalog No. 5000493082
Encompass_Preferred
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.
Catalog No. 50-004-93082 Supplier Chemglass Life Sciences Supplier No. CG1991V125
Suitable for reaction vials, often used in chemistry and pharmaceutical applications for compound synthesis, extraction, or sample preparation.