Learn More
Chemglass Life Sciences ECODryBlock for 25mm Round Bottom Tubes, 26.2mm O.D x 48.4mm Deep Well, 6 Wells
SDP
Catalog No. 5000493067
Encompass_Preferred
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.
Catalog No. 50-004-93067 Supplier Chemglass Life Sciences Supplier No. CG1991V107
Commonly used for general laboratory heating tasks, such as preparing solutions, dissolving substances, or incubating samples in routine chemical and biological analyses.