Learn More
Chemglass Life Sciences ECODryBlock for 42mm Round Bottom COD Test Tube, 42mm O.D x 51mm Deep Well, 4 Wells
SDP
Catalog No. 5000493114
Encompass_Preferred
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Catalog No. 50-004-93114 Supplier Chemglass Life Sciences Supplier No. CG1991V133
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.