Learn More
Chemglass Life Sciences ECODryBlock for 50mL Conical Bottom Centrifuge Tubes, 29mm O.D x 47.6mm Deep Well, 5 Wells
SDP
Catalog No. 5000493079
Encompass_Preferred
Used for medium to large capacity centrifugations, ideal for separating cells, particles, or biological components, as well as for preparing and collecting pellets.
Catalog No. 50-004-93079 Supplier Chemglass Life Sciences Supplier No. CG1991V115
Used for medium to large capacity centrifugations, ideal for separating cells, particles, or biological components, as well as for preparing and collecting pellets.