Learn More
Chemglass Life Sciences ECODryBlock Combo Block for 0.5ml, 1.5mL & 2.0ml Microcentrifuge Tube, 7.9mm O.D x 27.6mm Deep Well, 6 Wells & 11.1mm O.D x 39.1mm Deep Well, 10 Wells & 11.5mm O.D x 38.1mm Deep Well, 5 Wells
SDP
Catalog No. 5000493080
Encompass_Preferred
Provide maximum flexibility, allowing simultaneous heating of different tube formats, ideal for multipurpose laboratories handling diverse workflows.
Catalog No. 50-004-93080 Supplier Chemglass Life Sciences Supplier No. CG1991V118
Provide maximum flexibility, allowing simultaneous heating of different tube formats, ideal for multipurpose laboratories handling diverse workflows.