Learn More
Chemglass Life Sciences ECODryBlock Combo Block for 1.5ml, 15mL & 50ml Conical Centrifuge Tube, 11.1mm O.D x 39.1mm Deep Well, 4 Wells & 17.1mm O.D x 44.5mm Deep Well, 3 Wells & 29mm O.D x 47.6mm Deep Well, 2 Wells
SDP
Catalog No. 5000493104
Encompass_Preferred
Provide maximum flexibility, allowing simultaneous heating of different tube formats, ideal for multipurpose laboratories handling diverse workflows.
Catalog No. 50-004-93104 Supplier Chemglass Life Sciences Supplier No. CG1991V117
Provide maximum flexibility, allowing simultaneous heating of different tube formats, ideal for multipurpose laboratories handling diverse workflows.