Learn More
Chemglass Life Sciences ECODryBlock Combo Block for 16mm & 22mm Round Bottom COD Test Tubes, 16mm O.D x 47mm Deep Well, 18 Wells & 22mm O.D x 45.5 Deep Well, 2 Wells
SDP
Catalog No. 5000493094
Encompass_Preferred
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Catalog No. 50-004-93094 Supplier Chemglass Life Sciences Supplier No. CG1991V134
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.