Learn More
Velp Scientific ECODryBlock, Dry Block Heater

Description
- ControlPad with touchscreen and chemical-resistant surface
- One ControlPad controls up to two heating modules
- Each module holds two aluminum blocks (95 x 76 x 51 mm)
- Independent temperature control up to 165°C per module
- Independent timers per module
- Interchangeable aluminum blocks for different tubes, vials, and cuvettes
- Optional transparent lid reduces heat loss and improves temperature uniformity
- Supports external Pt100 and Pt1000 probes for direct temperature monitoring
Applications
- DNA/RNA extraction and purification
- Enzymatic reactions
- Cell culture activation
- DNA and protein denaturation
- Assay preparation and clinical testing
- Melting and boiling point determination
- Culture media testing
- General analytical protocols
Accessory
- Requires aluminum block(s) for operation
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Temperature Range (Metric) | 5°C to 165°C |
| Type | Dry Block Heater |
| No. of Blocks | 2 (sold separately) |
| Temperature Accuracy | ±0.5°C |
| Temperature Stability | ±0.2°C at 37°C, ±0.4 at 60°C, ±0.5°C at 165°C |
| Warranty | 3 Year |
| Dimensions (L x W x H) | 228 x 350 x 92.5 mm |
| Weight (English) | 4.6 lb. |
| Weight (Metric) | 2.1 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.