Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Velp Scientific ECODryBlock, Dry Block Heater - COD Analysis 4 Positions
SDP

Catalog No. 11392262
Change view
Click to view available options
Temperature Range (Metric):
5°C to 165°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
11-392-262 5°C to 165°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-392-262 Supplier Velp Scientific Supplier No. SC101A0700
Add to Cart
Add to Cart

Modular, digital, and precise dry block heater designed for sample preparation, incubation, and COD analysis.

  • ControlPad with 7 in. touchscreen and chemical-resistant surface
  • One ControlPad controls up to two heating modules
  • Each module holds two aluminum blocks (95 x 76 x 51 mm)
  • Independent temperature control up to 165°C per module
  • Independent timers per module
  • Interchangeable aluminum blocks for tubes, vials, and cuvettes
  • Optional transparent lid reduces heat loss and improves temperature uniformity
  • Supports external Pt100 and Pt1000 probes for direct temperature monitoring

Applications

  • COD analysis in water samples
  • DNA/RNA extraction and purification
  • Enzymatic reactions
  • Cell culture activation
  • DNA and protein denaturation
  • Assay preparation and clinical testing
  • Melting and boiling point determination
  • Culture media testing
  • General analytical protocols

Accessories included

  • COD-W04-D420-A block (cod. 31703693)

Accessories required for COD analysis

  • COD test tubes Ø 42 x 200 mm (3 pcs/box) (cod. 17129170)
  • Sample rack for COD-W04-D420-A block (cod. 31703702)
  • Air condenser with ground cone (cod. 11761455)
  • PTFE sheaths for 29/32 cones (cod. 15329634)
  • Antisplash bell (cod. 15349634)

Specifications

Autoclavable Not Autoclavable
Temperature Range (Metric) 5°C to 165°C
Type Dry Block Heater
Includes COD-W04-D420-A Block
Temperature Accuracy ±0.5°C
Temperature Stability ±0.5°C at 165°C
Warranty 3 Year
Dimensions (L x W x H) 228 x 350 x 92.5 mm
Weight (English) 4.6 lb.
Weight (Metric) 2.1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.