Learn More
Chemglass Life Sciences ECODryBlock, No ControlPad, 115-230V
SDP
Catalog No. 5000493085
Encompass_Preferred
ECODryBlock No ControlPad 115-230V. Modular digital dry block heater designed for accurate sample heating. Modular digital dry block heater designed for sample preparation incubation DNA denaturation enzymatic reactions culture media tests and COD analysis. ControlPad manages up to two heating Modules with space for up to 2 small aluminum blocks 76 x 51 x 95mm or 1 big aluminum block 152 x 51 x 95mm. Set independent timers and temperatures up to 165°C on each Module. Wide range of interchangeable aluminum blocks to accommodate different types of vials tubes and cuvette. Transparent cover creates warm air jacket that minimizes heat loss and ensures uniform temperature distribution. External probes Pt100 / Pt1000 enable precise monitoring directly within the block or the sample vessel. Possibility to set different temperatures on each Module. 7’’ capacitive touch screen w/ chemical-resistant glass surface. 3-year warranty. CG-1991-V-01X requires CG-1991-V-135
Catalog No. 50-004-93085 Supplier Chemglass Life Sciences Supplier No. CG1991V01X
ECODryBlock No ControlPad 115-230V. Modular digital dry block heater designed for accurate sample heating. Modular digital dry block heater designed for sample preparation incubation DNA denaturation enzymatic reactions culture media tests and COD analysis. ControlPad manages up to two heating Modules with space for up to 2 small aluminum blocks 76 x 51 x 95mm or 1 big aluminum block 152 x 51 x 95mm. Set independent timers and temperatures up to 165°C on each Module. Wide range of interchangeable aluminum blocks to accommodate different types of vials tubes and cuvette. Transparent cover creates warm air jacket that minimizes heat loss and ensures uniform temperature distribution. External probes Pt100 / Pt1000 enable precise monitoring directly within the block or the sample vessel. Possibility to set different temperatures on each Module. 7’’ capacitive touch screen w/ chemical-resistant glass surface. 3-year warranty. CG-1991-V-01X requires CG-1991-V-135