Learn More
New England Biolabs, Inc. EcoGII Methyltransferase – 200 units
SDP
Catalog No. NC1380830
Encompass
EcoGII Methyltransferase is a non-specific methyltransferase that modifies adenine residues (N6) in any sequence context.
Catalog No. NC1380830 Supplier New England Biolabs, Inc. Supplier No. M0603S
EcoGII Methyltransferase is a non-specific methyltransferase that modifies adenine residues (N6) in any sequence context.