Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ECOL(TM)

Catalog No. 1550125KGR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-501-25KGR 25 kg
15-501-1KGR 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-501-25KGR Supplier Sigma Aldrich Supplier No. 1550125KGR
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ECOL(TM)

Specifications

CAS 55-55-0
Melting Point 260°C (lit.)
Molecular Formula C7H9NO -+ 0.5H2SO4
Linear Formula C7H9NO -+ 1/2H2SO4
MDL Number MFCD00013140
Quantity 25 kg
Synonym 4-(Methylamino)phenol hemisulfate salt; Metol
Molecular Weight (g/mol) 172.19
Percent Purity ≥98% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.