Learn More
3B Scientific™ EcoLabBox Teaching Box, Environment (German)
SDPSCI_ED
Catalog No. S20760 Shop All 3B Scientific Products
Change view
Click to view available options
EcoLabBox Teaching Box, Environment (German)
Catalog No.
S20760 EcoLabBox Teaching Box, Environment (German)
1 options
Catalog No. S20760 Supplier 3B Scientific™ Supplier No. W11712
3B Scientific™ EcoLabBox Teaching Box, Environment is a veritable mobile laboratory for carrying out water, soil and air experiments out in the field. Detect and measure important substances affecting the environment
  • 21 fundamental experiments concerning the environment
  • Suitable for groups aged 12 and above
  • All reagents conform to German water hazard
  • An adjustable carrying strap for use when out and about
Specifications
Length (Metric) 38 cm
Width (Metric) 30 cm
Weight (Metric) 2.51 kg
Material Plastic
Includes Testing Equipment, Case
Product Type Biology Supplies