Learn More
Cederlane ECOLI ANTI POOL 1
SDP
Catalog No. NC9798456
Encompass
OK O Pool 1 EPEC/VTEC/STEC,(No. of tests: 150),(SSI� E. COLI OK O POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial
Catalog No. NC9798456 Supplier Cederlane Supplier No. 44292(SS)
OK O Pool 1 EPEC/VTEC/STEC,(No. of tests: 150),(SSI� E. COLI OK O POOL ANTISERA FOR SLIDE AGGLUTINATION), 3ml vial