Learn More
Cygnus Technologies ECOLI HCP ELISA KIT
Supplier:  Cygnus Technologies F410
Encompass
This kit was developed using broadly reactive polyclonal antibodies to the hundreds of different HCPs and is intended for use in determining the presence of E.coli host cell protein contamination in products manufactured by recombinant expression. Our assay validation has determined that the LOD for
Catalog No. NC9419378