Learn More
Cayman Chemical EcolI PhagocytosIs asy KI
SDP
Catalog No. 502892585
Encompass_Preferred
Cayman’s E. coli Phagocytosis Assay Kit employs FITC-labeled, heat-inactivated E. coli cells for the measurement of the phagocytic process in vitro. The engulfed fluorescent E. coli can be detected by flow cytometry.
Catalog No. 50-289-2585 Supplier Cayman Chemical Supplier No. 601370100 TEST
Cayman’s E. coli Phagocytosis Assay Kit employs FITC-labeled, heat-inactivated E. coli cells for the measurement of the phagocytic process in vitro. The engulfed fluorescent E. coli can be detected by flow cytometry.