Learn More
Crystal Chem Inc E Coli 360-HCP ELISA Starter Set (Includes types A, B, C, & D), 96-wells per type
SDP
Supplier:  Crystal Chem Inc IP2001
Encompass
E Coli 360-HCP ELISA Kit Starter Set. Includes 4 complete ELISA kits, one of each type. Crystal Chem, in partnership with German manufacturer BioGenes, has recently launched a new line of generic HCP ELISA kits for E Coli cells which far outperform competitor kits for detection of host cell proteins. Unlike traditional one-size-fits-all approaches, we provide four assays, each using a different anti-HCP antibody, so you can try multiple antibodies to find the one that works best for your process. 360-HCP ELISA has demonstrated better HCP recovery, better dilution linearity, and higher inter-assay precision than competitor’s kits.
Catalog No. 50-194-7939