Learn More
International Crystal ECON CARDS 95MM 13 MM PELLETS
Catalog No. NC9259033
Encompass_Preferred
Econo Card, 9.5mm, Ideal vehicle for mounting KBr pellets and polymer films or for use with disposable polyethylene IR transmission windows, They are available with 19mm and 9.5mm circular apertures. 100/PK
Catalog No. NC9259033 Supplier International Crystal Supplier No. 00122478
Econo Card, 9.5mm, Ideal vehicle for mounting KBr pellets and polymer films or for use with disposable polyethylene IR transmission windows, They are available with 19mm and 9.5mm circular apertures. 100/PK