Learn More
Ted Pella Inc Large Plain Economy Glass Microscope Slides, Beveled Corners, 3 x 2", 144/pk
SDP
Catalog No. NC0277576
Encompass
Made from clear soda lime corrosion resistant glass. Precleaned slides with a size of 3 x 2" and a 0.039 - 0.047" thickness (76.2 x 50.8mm and a 1-1.2mm thickness). Plain, ground edges and beveled corners, 144 slides per package.
Catalog No. NC0277576 Supplier Ted Pella Inc Supplier No. 260439
Made from clear soda lime corrosion resistant glass. Precleaned slides with a size of 3 x 2" and a 0.039 - 0.047" thickness (76.2 x 50.8mm and a 1-1.2mm thickness). Plain, ground edges and beveled corners, 144 slides per package.