Learn More
Sklar Instruments Econo(TM) Dressing Forceps, Sklar, 6 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. BX/12.
SDP
Catalog No. NC0731224 Shop All Sklar Instruments Products
Encompass_Preferred
Econo(TM) Dressing Forceps, Sklar, 6 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. Econo(TM) Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. BX12.
Catalog No. NC0731224 Supplier Sklar Instruments Supplier No. 21731
Econo(TM) Dressing Forceps, Sklar, 6 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. Econo(TM) Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. BX12.