Learn More
Apexbio Technology LLC Econazole 27220-47-9 1g
SDP
Catalog No. 502240802
Encompass_Preferred
Econazole (CAS 27220-47-9) is an imidazole-derived antifungal agent targeting cytochrome P450-dependent enzymes It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal membrane integrity and cell growth Econazole exerts its biological activity primarily through inhibition of cytochrome P450-dependent enzymes In experimental models involving fungi-mediated dermal infections and other dermatological assays econazole demonstrates inhibitory activity with reported IC50 values generally in the micromolar range depending on the fungal strain and specific assay employed Based on these pharmacological properties econazole holds research potential in dermatological and microbiological applications including investigations of fungal pathogenic mechanisms cellular susceptibility and resistance pathways
This item is not returnable. View return policy
Catalog No. 50-224-0802 Supplier Apexbio Technology LLC Supplier No. C63881000
Econazole (CAS 27220-47-9) is an imidazole-derived antifungal agent targeting cytochrome P450-dependent enzymes It is designed to inhibit ergosterol biosynthesis thereby disrupting fungal membrane integrity and cell growth Econazole exerts its biological activity primarily through inhibition of cytochrome P450-dependent enzymes In experimental models involving fungi-mediated dermal infections and other dermatological assays econazole demonstrates inhibitory activity with reported IC50 values generally in the micromolar range depending on the fungal strain and specific assay employed Based on these pharmacological properties econazole holds research potential in dermatological and microbiological applications including investigations of fungal pathogenic mechanisms cellular susceptibility and resistance pathways