Learn More
Medchemexpress LLC Econazole nitrate | 24169-02-6 | 98.0% | 25 G
SDP
Catalog No. 5000405582
Encompass_Preferred
Econazole nitrate is an imidazole class antifungal medication that also exhibits antibacterial activity. It is commonly used as a cream to treat various skin infections and has shown effectiveness in curing vaginal candidiasis via oral administration, with studies indicating it is generally well-tolerated.
  • Imidazole class antifungal medication
  • Exhibits antibacterial activity
  • Treats skin infections such as athlete's foot, tinea, pityriasis versicolor, ringworm, and jock itch
  • Cures vaginal candidiasis
  • Generally well tolerated
Catalog No. 50-004-05582 Supplier Medchemexpress LLC Supplier No. 25GHYB0453
Econazole nitrate is an imidazole class antifungal medication that also exhibits antibacterial activity. It is commonly used as a cream to treat various skin infections and has shown effectiveness in curing vaginal candidiasis via oral administration, with studies indicating it is generally well-tolerated.
  • Imidazole class antifungal medication
  • Exhibits antibacterial activity
  • Treats skin infections such as athlete's foot, tinea, pityriasis versicolor, ringworm, and jock itch
  • Cures vaginal candidiasis
  • Generally well tolerated