Learn More
Cayman Chemical Econazole (nitrate), Purity: ≥98%, CAS: 24169-02-6, 5 g
SDP
Catalog No. 502984995
Encompass_Preferred
An antifungal compound that acts by inhibiting the 14α-demethylation of lanosterol, which consequently leads to the inhibition of ergosterol synthesis in fungal cell membranes
Catalog No. 50-298-4995 Supplier Cayman Chemical Supplier No. 202235 G
An antifungal compound that acts by inhibiting the 14α-demethylation of lanosterol, which consequently leads to the inhibition of ergosterol synthesis in fungal cell membranes