Learn More
New England Biolabs, Inc. EcoNI – 1000 units
SDP
Catalog No. 50812174
Encompass
A restriction endonuclease that recognizes the sequence CCTNN^N_NNAGG.
Catalog No. 50-812-174 Supplier New England Biolabs, Inc. Supplier No. R0521S
A restriction endonuclease that recognizes the sequence CCTNN^N_NNAGG.