Learn More
New England Biolabs, Inc. EcoNI – 5000 units
SDP
Catalog No. 50812173
Encompass
A restriction endonuclease that recognizes the sequence CCTNN^N_NNAGG.
Catalog No. 50-812-173 Supplier New England Biolabs, Inc. Supplier No. R0521L
A restriction endonuclease that recognizes the sequence CCTNN^N_NNAGG.