Learn More
Bio Rad Laboratories ECONO-COLUMN CHROMATOGRAPHY CO
Catalog No. 509012356
Encompass
Column, Chromatography; Bio-Rad; Econo-Column; Autoclavable; Maximum operating pressure <1 bar (14.7 psi); 0.7 x 15cm; 6mL; Quantity: 2
Catalog No. 50-901-2356 Supplier Bio Rad Laboratories Supplier No. 7370717
Column, Chromatography; Bio-Rad; Econo-Column; Autoclavable; Maximum operating pressure <1 bar (14.7 psi); 0.7 x 15cm; 6mL; Quantity: 2