Learn More
Bio Rad Laboratories ECONO-COLUMN CHROMATOGRAPHY CO
Supplier:  Bio Rad Laboratories 7371032
Encompass
Column, Chromatography; Bio-Rad; Econo-Column; Autoclavable; Maximum operating pressure <1 bar (14.7 psi); 1.0 x 30cm; 24mL; Quantity: 2
Catalog No. 50-901-2482