Learn More
Bio Rad Laboratories ECONO-COLUMN CHROMATOGRAPHY CO
Supplier:  Bio Rad Laboratories 7371512
Encompass
Column, Chromatography; Bio-Rad; Econo-Column; Glass; Autoclavable; Operates at pressure of <1 atm (14.7psi); Accepts both Econo-Column funnels and Econo-Column flow adaptors; 1.5 x 10cm; 18mL; 2
Catalog No. 50-901-0024