Learn More
Bio Rad Laboratories ECONO-COLUMN CHROMATOGRAPHY CO
Supplier:  Bio Rad Laboratories 7372593
Encompass
Column, Chromatography; Bio-Rad; Econo-Column; Autoclavable; Maximum operating pressure <1 bar (14.7 psi); 2.5 x 120cm; 589mL; Quantity: 2
Catalog No. 50-901-2367