Learn More
Sklar Instruments Econo Crile Forceps, Sklar, 5-1/2 in., Straight, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437636 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Crile Forceps, Sklar, 5-1/2 in., Straight, Serrated, Stainless Steel. Econo Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. There is a ratchet locking mechanism for a tight, secure hold on vessels. They may also be used for soft tissue dissection typically no deeper than 6-8 inches or in Laparotomy procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 5-1/2 inches. It is sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7636 Supplier Sklar Instruments Supplier No. 21447
Econo Crile Forceps, Sklar, 5-1/2 in., Straight, Serrated, Stainless Steel. Econo Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. There is a ratchet locking mechanism for a tight, secure hold on vessels. They may also be used for soft tissue dissection typically no deeper than 6-8 inches or in Laparotomy procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 5-1/2 inches. It is sold in boxes of 12.