Learn More
Sklar Instruments Econo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. PK/12.
SDP
Catalog No. 501437604 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. The straight version is typically used for cutting surface tissue or sutures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. 50-143-7604 Supplier Sklar Instruments Supplier No. 21327
Econo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Stainless Steel. Econo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. The straight version is typically used for cutting surface tissue or sutures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.