Learn More
Sklar Instruments Econo Dressing Forceps, Sklar, 4-1/2 in., Blunt, Dressing Forceps, Straight, Serrated, Stainless Steel. PK/12.
SDP
Catalog No. 501437674 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Dressing Forceps, Sklar, 4-1/2 in., Blunt, Dressing Forceps, Straight, Serrated, Stainless Steel. Econo Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 4-1/2 inches. They are sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7674 Supplier Sklar Instruments Supplier No. 21705
Econo Dressing Forceps, Sklar, 4-1/2 in., Blunt, Dressing Forceps, Straight, Serrated, Stainless Steel. Econo Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with serrated tips and a length of 4-1/2 inches. They are sold in boxes of 12.