Learn More
Bevco Precision Manufacturing Co Econo ESD Chair with Casters; Desk
Catalog No. NC2398365 Shop All Bevco Precision Manufacturing Co Products
Encompass_Preferred
Bevco's Lexington ESD Series chairs deliver all of the outstanding ergonomic benefits at a lower price point. This reliable chair was built to meet the demands of daily use for environments that require some level of static discharge control but do not need to meet ANSI/ESD 12.1 - 2019 specifications. Static dissipative with a resistance of 105-109 ohms when measured from seat/back to grounding chain only.Fully adjustable contoured back has a bumper guard on the edges to protect the upholstery.A waterfall seat aids lower body circulation promoting overall wellness.The pneumatic height adjustment allows you to easily change your seat height from 17" - 22".Upholstered fabric meets CAL 117 standards.The chair features a sturdy 25" diameter black nylon base with a brass drag chain. Model comes standard with dual-wheel hard floor casters.
Catalog No. NC2398365 Supplier Bevco Precision Manufacturing Co Supplier No. V800SHC
Bevco's Lexington ESD Series chairs deliver all of the outstanding ergonomic benefits at a lower price point. This reliable chair was built to meet the demands of daily use for environments that require some level of static discharge control but do not need to meet ANSI/ESD 12.1 - 2019 specifications. Static dissipative with a resistance of 105-109 ohms when measured from seat/back to grounding chain only.Fully adjustable contoured back has a bumper guard on the edges to protect the upholstery.A waterfall seat aids lower body circulation promoting overall wellness.The pneumatic height adjustment allows you to easily change your seat height from 17" - 22".Upholstered fabric meets CAL 117 standards.The chair features a sturdy 25" diameter black nylon base with a brass drag chain. Model comes standard with dual-wheel hard floor casters.