Learn More
Research Products International Corp EKONO Hybridization Buffer, 1 Liter
SDP
Catalog No. 50212604
Encompass
EKONO provides real cost savings without sacrificing performance or reliability. Effective for Southern, Northern, Southwestern and oligonucleotide hybridizaiton. Can be used with radioactive and non-radioactive probes or for screening libraires. Simple overnight incubation produces high sensitivity with minimum background.
Supplied in a 1 liter bottle.
Store at room temperature.
Catalog No. 50-212-604 Supplier Research Products International Corp Supplier No. 248800
EKONO provides real cost savings without sacrificing performance or reliability. Effective for Southern, Northern, Southwestern and oligonucleotide hybridizaiton. Can be used with radioactive and non-radioactive probes or for screening libraires. Simple overnight incubation produces high sensitivity with minimum background.
Supplied in a 1 liter bottle.
Store at room temperature.