Learn More
Sklar Instruments Econo Iris Scissors, 4-1/2 in., Straight, Sharp/Sharp, Smooth, Finger Rings, BX of 12.
SDP
Catalog No. 501109749 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Iris Scissors, 4-1/2 in., Straight, Sharp/Sharp, Smooth, Finger Rings, BX of 12. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth edges, sharp tips, and a length of 4-1/2 inches. They are available in a box of 12.
Catalog No. 50-110-9749 Supplier Sklar Instruments Supplier No. 21104
Econo Iris Scissors, 4-1/2 in., Straight, Sharp/Sharp, Smooth, Finger Rings, BX of 12. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is straight with smooth edges, sharp tips, and a length of 4-1/2 inches. They are available in a box of 12.