Learn More
Bio Rad Laboratories ECONO-PAC CHROMATOGRAPHY COLUM
Catalog No. 509011607
Encompass
Column, Chromatography Econo-Pac Bio-Rad Empty Includes upper bed support, end cap, tip closure 14cm high 1.5 x 12cm PP 10mL reservoir Graduated 1-20mL bed vol. Snap-off tip Autoclavable 50
Catalog No. 50-901-1607 Supplier Bio Rad Laboratories Supplier No. 7321010
Column, Chromatography Econo-Pac Bio-Rad Empty Includes upper bed support, end cap, tip closure 14cm high 1.5 x 12cm PP 10mL reservoir Graduated 1-20mL bed vol. Snap-off tip Autoclavable 50