Learn More
MSC Weiler Cleaning and Finishing Brush, Synthetic, Foam, Black, 1 ea
Catalog No. 502867552
Encompass
This cleaning and finishing brush features synthetic bristles and a foam handle, designed for effective cleaning and finishing tasks. It has a brush length of 11-3/16 in and a brush width of 15/16 in.
  • Synthetic bristle material
  • Foam handle for comfortable grip
  • Black bristle color
  • Brush length measures 11-3/16 in
  • Brush width measures 15/16 in
  • Bristle length is 2.5 in
  • Not flagged
Catalog No. 50-286-7552 Supplier MSC Supplier No. 75851899
This cleaning and finishing brush features synthetic bristles and a foam handle, designed for effective cleaning and finishing tasks. It has a brush length of 11-3/16 in and a brush width of 15/16 in.
  • Synthetic bristle material
  • Foam handle for comfortable grip
  • Black bristle color
  • Brush length measures 11-3/16 in
  • Brush width measures 15/16 in
  • Bristle length is 2.5 in
  • Not flagged