Learn More
Sklar Instruments Econo Scalpel Handle, Sklar, 6-1/2 in., Straight, Stainless Steel. #7 PK/12.
SDP
Catalog No. 501437520 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Scalpel Handle, Sklar, 6-1/2 in., Straight, Stainless Steel. Econo #7 Scalpel Handle has a thin shank used for fine, precise incisions made in a small space or deep wound. This handle holds size 10 series disposable blades (10, 11, 12, 15.) Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. 50-143-7520 Supplier Sklar Instruments Supplier No. 21042
 Restricted Item
Add to Cart
Add to Cart
Econo Scalpel Handle, Sklar, 6-1/2 in., Straight, Stainless Steel. Econo #7 Scalpel Handle has a thin shank used for fine, precise incisions made in a small space or deep wound. This handle holds size 10 series disposable blades (10, 11, 12, 15.) Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.