Learn More
Sklar Instruments Econo Scalpel Handle, Sklar, 5 in., Straight, Stainless Steel. Econo Scalpel Handle #3 PK/12.
SDP
Catalog No. 501437518 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Scalpel Handle, Sklar, 5 in., Straight, Stainless Steel. Econo Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. This #3 handle has centimeter and millimeter markings on the handle for measuring tissue. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. 50-143-7518 Supplier Sklar Instruments Supplier No. 21035
Econo Scalpel Handle, Sklar, 5 in., Straight, Stainless Steel. Econo Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. This #3 handle has centimeter and millimeter markings on the handle for measuring tissue. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.