Learn More
Sklar Instruments Econo Multi-Cut Utility Scissors, Sklar, Lime Green, 7in., Blunt/Blunt, Angled, Serrated, Finger Rings / 1 Large Ring, Stainless Steel / Plastic. Box of 12.
SDP
Catalog No. 501437529 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Multi-Cut Utility Scissors, Sklar, Lime Green, 7 in., Blunt/Blunt, Angled, Serrated, Finger Rings / 1 Large Ring, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This item is sold in boxes of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7529 Supplier Sklar Instruments Supplier No. 21105LG
Econo Multi-Cut Utility Scissors, Sklar, Lime Green, 7 in., Blunt/Blunt, Angled, Serrated, Finger Rings / 1 Large Ring, Stainless Steel / Plastic. Econo Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes. These scissors are angled and have one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This item is sold in boxes of 12.