Learn More
Sklar Instruments Single Use, Econo-Sterile, Splinter Forceps, Sklar 4-1/2 in., Straight, Serrated, Stainless Steel, Disposable.CS of 50.
SDP
Catalog No. 501094222 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Splinter Forceps, Sklar. 4-1/2 in., Straight, Serrated, Stainless Steel, Disposable.CS of 50. Sterile Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects.
Catalog No. 50-109-4222 Supplier Sklar Instruments Supplier No. 962410
Single Use, Econo-Sterile, Splinter Forceps, Sklar. 4-1/2 in., Straight, Serrated, Stainless Steel, Disposable.CS of 50. Sterile Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects.