Learn More
Bio Rad Laboratories Econo-Column Chromatography Columns | 2/PK
Supplier:  Bio Rad Laboratories 7372507
Encompass
Use these empty glass chromatography columns with low-pressure (<1 bar) chromatography systems or gravity flow separations. Econo-Column columns are ideal for simple packing and procedure running. They are 2.5x5cm with a 25ml volume. The porous polymerbed support at the bottom of the column retains fine particles. They have translucent polypropylene end fittings which allow viewing of the entire column bed. The columns are autoclavable and NaOH compatible. Includes one stopcock per column ordered.
Catalog No. NC1860165