Learn More
Sklar Instruments Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. Econo instruments are sold in bulk quantities.
SDP
Catalog No. 501437517 Shop All Sklar Instruments Products
Encompass_Preferred
Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.
This item is not returnable. View return policy
Catalog No. 50-143-7517 Supplier Sklar Instruments Supplier No. 21033
Econo Surgical Knife Handle, Sklar, Straight, Stainless Steel. Econo Scalpel Handle #3 is compatible with size 10 series disposable blades (10, 11, 12, 15) and used for sharp dissection. This handle has horizontal serrations for thumb and forefinger gripping. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities.