Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Economical Leather Palm Work Gloves - Gauntlet Cuff

Catalog No. 19148747 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-148-747 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-148-747 Supplier Wells Lamont™ Supplier No. Y3409L
This item is not returnable. View return policy

Gloves feature a standard shoulder split leather palm, wing thumb, and gauntlet cuff.

  • Economical protection
  • Standard shoulder split leather
  • Gauntlet cuff
  • Wing thumb

Specifications

Cuff Material Fabric
Color Gray
Grade Economy
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type General Purpose Gloves
Size Large
Glove Cuff Color Blue with Red Stripes
Glove Cuff Style Gauntlet
Hem Color Brown
Series Y3409
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.