Learn More
Sigma Aldrich Fine Chemicals Biosciences Economical neoprene dry-bo
Catalog No. 502901684
Encompass
Neoprene bonded sleeve and glove.General purpose elastomer. Resists moderate chemicals and acids ozone oils fats greases many oils and solvents. Attacked by strong oxidizing acids esters ketones chlorinated aromatic and nitro hydrocarbons.
Catalog No. 50-290-1684 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1700621PAK
Neoprene bonded sleeve and glove.General purpose elastomer. Resists moderate chemicals and acids ozone oils fats greases many oils and solvents. Attacked by strong oxidizing acids esters ketones chlorinated aromatic and nitro hydrocarbons.