Learn More
Sigma Aldrich Fine Chemicals Biosciences Economical neoprene dry-bo
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1700701PAK
Encompass
Neoprene bonded sleeve and glove.General purpose elastomer. Resists moderate chemicals and acids ozone oils fats greases many oils and solvents. Attacked by strong oxidizing acids esters ketones chlorinated aromatic and nitro hydrocarbons.
Catalog No. 50-290-1708