Learn More
Bioworld Economou & Read Basal Medium, 25 L
Catalog No. 50254899
Encompass_Preferred
Economou & Read Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Economou & Read (1984)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.84 g/LpH: ≈4.5Soluble in water
Catalog No. 50-254-899 Supplier Bioworld Supplier No. 306301622
Economou & Read Basal MediumContains Macronutrients, Micronutrients, and Vitamins as described by Economou & Read (1984)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 1.84 g/LpH: ≈4.5Soluble in water